missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA7L Monoclonal Antibody (OTI4A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803021
This item is not returnable.
View return policy
Description
Cell division cycle-associated 7-like protein (or RAM2) is a transcription factor which plays a role in transcriptional regulation and apoptotic signaling pathways. It acts as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.Specifications
| CDCA7L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GN5 | |
| CDCA7L | |
| Human recombit protein fragment corresponding to amino acids 1-337 of human CDCA7L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4A2 | |
| Unconjugated | |
| CDCA7L | |
| JPO2, R1, RAM2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55536 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction