missing translation for 'onlineSavingsMsg'
Learn More

Cdk4 Monoclonal Antibody (DCS-31), Invitrogen™

Product Code. 17727587
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17727587 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17727587 Supplier Invitrogen Supplier No. MA513498

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifically detect Cdk4 in Human, Mouse, Rat samples using Thermo Scientifc™ Pierce™ anti-Cdk4 Clone: DCS-31, validated for FACS, IP and WB

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Entrez Gene ID:94201

Synonyms:cell division kinase 4, cell division protein kinase 4, melanoma cutaneous malignant, 3

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk4
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-31
Conjugate Unconjugated
Gene Accession No. P35426
Gene Symbols Cdk4
Host Species Mouse
Immunogen Recombinant cdk4 protein
Quantity 500 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.