missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk6 Antibody (8G3), Novus Biologicals™
Rabbit Monoclonal Antibody
355.00 € - 533.00 €
Specifications
| Antigen | Cdk6 |
|---|---|
| Clone | 8G3 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043120
|
Novus Biologicals
NBP3-26663-50ul |
50 μL |
355.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043198
|
Novus Biologicals
NBP3-26663-100ul |
100 μL |
533.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Cdk6 | |
| Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE | |
| A synthesized peptide derived from Human Cdk6 [UniProt Q00534] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8G3 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1021 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title