missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL5 Antibody (CL4881) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61145C
This item is not returnable.
View return policy
Description
CDKL5 Monoclonal antibody specifically detects CDKL5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CDKL5 | |
| Monoclonal | |
| DyLight 650 | |
| cyclin-dependent kinase-like 5, serine/threonine kinase 9, serine/threonine-protein kinase 9 | |
| This antibody was developed using a recombinant protein derived from O76039, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Protein Kinase | |
| 6792 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4881 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction