missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDP/CUTL1 Antibody (CL5275) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61408PE
This item is not returnable.
View return policy
Description
CDP/CUTL1 Monoclonal antibody specifically detects CDP/CUTL1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CDP/CUTL1 | |
| Monoclonal | |
| PE | |
| CCAAT displacement protein, CDP/Cux, CDPCCAAT displacement protein, COY1, CUT, cut (Drosophila)-like 1 (CCAAT displacement protein), cut-like 1, CCAAT displacement protein (Drosophila), cut-like homeobox 1, FLJ31745, golgi integral membrane protein 6, GOLIM6, homeobox protein cut-like 1, Homeobox protein cux-1, Nbla10317, p100, p110, p200, p75, protein CASP, putative protein product of Nbla10317 | |
| This antibody was developed using a recombinant protein derived from P39880, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5275 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1523 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction