missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDSN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13184-1-AP-20UL
This item is not returnable.
View return policy
Description
CDSN, also named as S protein, is important for the epidermal barrier integrity. CDSN is a unique extracellular 52- to 56- kDa glycoprotein found in corneodesmosomes. Pathogenic mutations in CDSN have been linked in humans to genodermatoses affecting both the hair follicle and the epidermis. Inflammatory peeling skin syndrome is caused a novel mutation in CDSN.Specifications
| CDSN | |
| Polyclonal | |
| Unconjugated | |
| CDSN | |
| CDSN, corneodesmosin, D6S586E, HTSS, S | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1041, 386463 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15517, Q7TPC1 | |
| Cdsn | |
| CDSN Fusion Protein Ag3854 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction