missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdx1 Antibody (123a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
201.00 € - 351.00 €
Specifications
| Antigen | Cdx1 |
|---|---|
| Clone | 123a |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing |
| Applications | Western Blot, Immunofluorescence, ChIP Assay, ChIP sequencing (ChIP-seq) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658088
|
Novus Biologicals
NBP2-50160-0.025ml |
0.025 mL |
201.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602278
|
Novus Biologicals
NBP2-50160 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdx1 Monoclonal antibody specifically detects Cdx1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), ChIP assay SequencingSpecifications
| Cdx1 | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence, ChIP Assay, ChIP sequencing (ChIP-seq) | |
| Unconjugated | |
| Mouse | |
| Epigenetics | |
| PBS | |
| 1044 | |
| IgG1 | |
| Protein A purified |
| 123a | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
| Human Cdx1 N-terminal peptide | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title