missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (CDX2/9298R), Novus Biologicals™
Mouse Monoclonal Antibody
245.00 € - 486.00 €
Specifications
| Antigen | CDX2 |
|---|---|
| Clone | CDX2/9298R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615334
|
Novus Biologicals
NBP3-23686-20ug |
20 μg |
245.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645064
|
Novus Biologicals
NBP3-23686-100ug |
100 μg |
486.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CDX2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 1045 | |
| IgG κ | |
| Protein A purified |
| CDX2/9298R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Recombinant full-length human CDX2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title