missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Antibody (CL12974) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18552MFV610
This item is not returnable.
View return policy
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CDX2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| This antibody was developed using a synthetic peptide derived from Q99626, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 1045 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL12974 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction