missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CEACAM1/CD66a Antibody (cergutuzumAb) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28550-50ug
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| Unconjugated | |
| P06731 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| cergutuzumAb | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
| CEACAM5/ CEA/ CD66e | |
| 50 μg | |
| Apoptosis, Cancer | |
| 634 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction