missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM20 Antibody (1032339) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9416JF525
This item is not returnable.
View return policy
Description
CEACAM20 Monoclonal antibody specifically detects CEACAM20 in Human samples. It is validated for Flow Cytometry
Specifications
| CEACAM20 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| UNQ9366 | |
| Human embryonic kidney cell HEK293-derived human CEACAM-20, Gln31-Gly450, Accession # Q6UY09-1 | |
| RUO | |
| 125931 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 1032339 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction