missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM20 Monoclonal Antibody (1032339)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562024
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Together with the tyrosine-protein kinase SYK, enhances production of the cytokine CXCL8/IL-8 via the NFKB pathway and may thus have a role in the intestinal immune response.
Specifications
| CEACAM20 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UY09 | |
| CEACAM20 | |
| Human embryonic kidney cell HEK293-derived human CEACAM-20, Gln31-Gly450. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 1032339 | |
| Unconjugated | |
| CEACAM20 | |
| 9130012D09Rik; carcinoembryonic antigen related cell adhesion molecule 20; carcinoembryonic antigen-related cell adhesion molecule 20; Ceacam20; CEA-related cell adhesion molecule 20; GPAD9366; UNQ9366; UNQ9366/PRO34155 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 125931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction