missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (C66/1291), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53350-20UG
This item is not returnable.
View return policy
Descrizione
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifica
| CEACAM5/CD66e | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Mouse | |
| 140 kDa | |
| 20 μg | |
| Cancer, Cellular Markers, Immunology | |
| 1048 | |
| Store at 4C. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| C66/1291 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| CEACAM5 | |
| Recombinant full-length human CEA protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido