missing translation for 'onlineSavingsMsg'
Learn More

Cefepime hydrochloride monohydrate

Product Code. 15487509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487509 1 g 1g
15497509 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487509 Supplier Thermo Scientific Alfa Aesar Supplier No. J66237.03
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cefepime hydrochloride monohydrate, CAS # 123171-59-5, is a hydrochloride salt of the antibiotic derived from Acremonium fungus. | CAS: 123171-59-5 | C19H29Cl2N6O6S2+ | 572.497 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria

• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis

Application

• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance

• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123171-59-5
Molecular Formula C19H29Cl2N6O6S2+
Molecular Weight (g/mol) 572.497
MDL Number MFCD01687679
InChI Key LRAJHPGSGBRUJN-OMIVUECESA-O
PubChem CID 129893984
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
SMILES C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl

Specifications

Type Cefepime Hydrochloride Monohydrate
Physical Form Powder
Melting Point 150°C
Color White to Yellow
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in DMSO
Formula Weight 571.5
Chemical Name or Material Cefepime hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.