missing translation for 'onlineSavingsMsg'
Learn More

Cefoperazone, MedChemExpress

Product Code. 16452089
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452089 1 g 1g
16420828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452089 Supplier Medchem Express Supplier No. HYB0210
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cefoperazone, a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.

Cefoperazone, a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.

Specifications

Chemical Name or Material Cefoperazone
Physical Form Solid
CAS 62893-19-0
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C25H27N9O8S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (154.88 mM) ∣H2O : 0.1 mg/mL (0.15 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(CSC3=NN=NN3C)CS[C@]2([H])[C@H](NC([C@H](NC(N4C(C(N(CC)CC4)=O)=O)=O)C5=CC=C(O)C=C5)=O)C1=O)O
Molecular Weight (g/mol) 645.67
Formula Weight 645.67
Percent Purity 99.82%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.