missing translation for 'onlineSavingsMsg'
Learn More

Melford Cefotaxime

Product Code. p-200087269
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018170 5 g 5g
30018169 25 g 25g
30018168 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018170 Supplier Melford Supplier No. C510005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cefotaxime Sodium Salt is a third-generation cephalosporin antibiotic.

Cefotaxime, as a cephalosporin, exerts its antibacterial effects by inhibiting bacterial cell wall synthesis. It does this by binding to penicillin-binding proteins (PBPs), disrupting the final step of peptidoglycan synthesis in the bacterial cell wall. This interference leads to structural instability and eventual lysis of the bacterial cell.

Cefotaxime sodium salt is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cefotaxime.

Specifications

Chemical Name or Material Cefotaxime
CAS 64485-93-4
Quantity 5 g
Molecular Formula C16H16N5NaO7S2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.