missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefotaxime, 98%

Product Code. 15535136 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15535136 1 g 1g
15482154 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535136 Supplier Thermo Scientific Chemicals Supplier No. 455270010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cefotaxime, CAS # 63527-52-6, is a beta-lactam antibiotic that has activity against gram-negative and positive bacteria

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cefotaxime, is an antibiotic with broad-spectrum activity against aerobic and anaerobic bacteria
  • It has been shown to have a high frequency of resistance mechanisms against strains such as Pseudomonas aeruginosa

Applications

  • L Cefotaxime has broad spectrum antibacterial activity
  • It binds to the penicillin-binding proteins (PBPs), resulting in inhibition of the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63527-52-6
Molecular Formula C16H17N5O7S2
Molecular Weight (g/mol) 455.46
InChI Key GPRBEKHLDVQUJE-YGRLFVJLSA-N
PubChem CID 53850837
IUPAC Name (6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)O

Specifications

Type Cefotaxime
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 455.47
Chemical Name or Material Cefotaxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.