missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOKU-E™ Cefotaxime ReadyMade™ Solution
Description
- CAS Number: 64485-93-4
- Molecular Formula: C16H16N5NaO7S2
- Molecular Weight: 447.45 g/mo
- Storage Conditions: -20°C
- Form: Liquid
- Source: Semi-synthetic (Acremonium sp.)
- Spectrum: Cefotaxime Sodium has broad-spectrum activity against a wide variety of Gram-positive and Gram-negative bacteria. However, unlike many cephalosporin antibiotics, Cefotaxime Sodium is not effective against Pseudomonas aeruginosa
- Mechanism of Action: Like β-lactams, cephalosporins interfere with PBP (penicillin binding protein) activity involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues providing additional strength to the cell wall. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised and ultimately leads to cell lysis and death. Resistance to cephalosporins is commonly due to cells containing plasmid encoded β-lactamases. Cefotaxime Sodium however, is largely resistant to β-lactamases
Specifications
Specifications
| Chemical Name or Material | Cefotaxime |
| Physical Form | Liquid |
| CAS | 64485-93-4 |
| Source | Semi-synthetic (Acremonium sp.) |
| Packaging | 2 mL plastic vial |
| Sterile Filtered | Yes, 0.22 μm |
| Sterility | Sterile |
| Color | Colorless |
| Concentration | 100 to 105 mg/mL |
| Quantity | 10 x 1 mL |
| Show More |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?