missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime Sodium Salt, ≥95%, MP Biomedicals™

Product Code. 11409101 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409101 500 mg 500mg
11499091 0.1 g 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409101 Supplier MP Biomedicals Supplier No. 0215494790
  • ONLINE EXCLUSIVE PRICE
    296.00 €
    280.35 € / 500mg
    Save 15.65 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 64485-93-4
Molecular Formula C16H16N5NaO7S2
Molecular Weight (g/mol) 477.44
MDL Number MFCD00079073
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
Synonym (6R-(6-α,7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4,2,0) oct-2-ene-2-carboxylic acid, sodium salt, (6R-(6-α, 7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4, 2
PubChem CID 88631411
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Cefotaxime Sodium Salt
Physical Form Powder
Color White
Quantity 500 mg
Assay Percent Range ≥95%
IUPAC Name sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Formula Weight 477.5
Chemical Name or Material Cefotaxime Sodium Salt
compliance-icons compliance-icons
Product Identifier
  • Cefotaxime Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cefotaxime Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.