missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime Sodium Salt, MP Biomedicals™

Product Code. 11419101 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419101 Supplier MP Biomedicals Supplier No. 0215494791
  • ONLINE EXCLUSIVE PRICE
    481.00 €
    455.70 € / 1g
    Save 25.30 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits bacterial cell wall synthesis. Cefotaxime Sodium Salt, MP Biomedicals is a cephalosporin antibiotic that is active against both gram-negative and gram-positive bacteria. It is used for infections of the respiratory tract, skin, bones, joints, uro

Cefotaxime binds to penicillin-binding proteins (PBPs), which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes. Cefotaxime blocks the division of cyanobacteria and lower plant organelles/plastids such as the photosynthetic organelles of Glaucophytes and chloroplasts of bryophytes. Cefotaxime is used to treat a variety of bacterial infections:

  • Good coverage against most Gram-negative bacteria, with the notable exception of Pseudomonas
  • Effective against most Gram-positive cocci, except for Enterococcus
  • Active against penicillin-resistant strains of Streptococcus pneumoniae
  • Modest activity against the anaerobic Bacteroides fragilis
  • In meningitis, cefotaxime crosses the blood–brain barrier better than cefuroxime

Specifications

Physical Form Powder
CAS 64485-93-4
Quantity 1 g
Molecular Formula C16H16N5NaO7S2
MDL Number MFCD00079073
Synonym Cefotaxime sodium, (6R-(6-a,7-b(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4,2,0) oct-2-ene-2-carboxylic acid, sodium salt
Solubility Information Soluble in water (0.05gm/1mL, clear, light yellow solution).
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
IUPAC Name sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 477.44
PubChem CID 88631411
Formula Weight 477.45
Percent Purity ≥95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cefotaxime Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cefotaxime Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.