Learn More
Melford Cefsulodin
Description
Cefsulodin Sodium Salt is a cephalosporin antibiotic. Cefsulodin inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
Effective against a range of Gram-negative bacteria, it is particularly useful in the isolation and selection of these bacteria in microbiological and research settings. Commonly used in culture media for the growth and identification of specific bacterial strains.
Additionally, it is often employed as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of Gram-negative bacteria lacking resistance to cefsulodin.
Specifications
Specifications
| Chemical Name or Material | Cefsulodin |
| CAS | 52152-93-9 |
| Quantity | 100 mg |
| Molecular Formula | C22H19N4NaO8S2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.