missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin sodium salt, 99% purity, For HPLC analysis, 93%, MP Biomedicals™

Product Code. 11456801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456801 0.1 g 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456801 Supplier MP Biomedicals Supplier No. 0219867780
  • ONLINE EXCLUSIVE PRICE
    102.00 €
    96.60 € / 100mg
    Save 5.40 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Off-white solid cephalosporin antibiotic.

Specifications

Type Cefsulodin Sodium Salt
Physical Form Crystalline Powder
CAS 52152-93-9
Color White
Quantity 0.1 g
Assay Percent Range 0.93
Molecular Formula C22H21N4NaO8S2+
InChI Key GTZPOHRNWUTXNB-DWBVFMGKSA-O
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]
IUPAC Name (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
Molecular Weight (g/mol) 556.54
PubChem CID 124203950
Formula Weight 554.5
Percent Purity 99%
Grade Not Available
Chemical Name or Material Cefsulodin sodium salt, For HPLC analysis
Show More Show Less
compliance-icons
Product Identifier
  • Cefsulodin Sodium Salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cefsulodin Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.