missing translation for 'onlineSavingsMsg'
Learn More

Ceftiofur sodium, 95%, Thermo Scientific Chemicals

Product Code. 17836354
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17836354 1 g 1g
15458429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17836354 Supplier Thermo Scientific Alfa Aesar Supplier No. J66886.03
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A broad spectrum antibiotic | CAS: 104010-37-9 | C19H16N5NaO7S3 | 545.53 g/mol

Ceftiofur sodium is resistant to the antibiotic resistance enzyme beta-lactamase, and has activity against Gram-positive and Gram-negative bacteria. E. coli strains resistant to ceftiofur have been reported. The bactericidal activity of ceftiofur results from the inhibition of cell wall synthesis via affinity for penicillin-binding proteins. The PBPs are transpeptidases which are vital in peptidoglycan biosynthesis. Therefore, their inhibition prevents this vital cell wall component from being properly synthesized.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ceftiofur sodium is resistant to the antibiotic resistance enzyme beta-lactamase, and has activity against Gram-positive and Gram-negative bacteria. E. coli strains resistant to ceftiofur have been reported. The bactericidal activity of ceftiofur results from the inhibition of cell wall synthesis via affinity for penicillin-binding proteins. The PBPs are transpeptidases which are vital in peptidoglycan biosynthesis. Therefore, their inhibition prevents this vital cell wall component from being properly synthesized.

Solubility
Freely soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104010-37-9
Molecular Formula C19H16N5NaO7S3
Molecular Weight (g/mol) 545.53
MDL Number MFCD01766184
InChI Key RFLHUYUQCKHUKS-JUODUXDSSA-M
PubChem CID 131632581
IUPAC Name sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Ceftiofur Sodium
Physical Form Powder
Color White or Yellow
Quantity 1 g
Solubility Information Freely soluble in aqueous solution
Formula Weight 545.54
Percent Purity 95%
Chemical Name or Material Ceftiofur sodium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.