missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Cefuroxime Sodium Salt
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Ceftriaxone Disodium Salt is a third-generation cephalosporin antibiotic. The hemi (heptahydrate) form indicates the presence of water molecules in the crystal structure.
Ceftriaxone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis. This interference leads to structural instability and eventual cell lysis. Broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. May be used in microbiology research and studies focusing on antibacterial agents. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.
Specifications
Specifications
| Chemical Name or Material | Cefuroxime, Sodium Salt |
| CAS | 56238-63-2 |
| Quantity | 1 g |
| Molecular Formula | C16H15N4NaO8S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction