missing translation for 'onlineSavingsMsg'
Learn More

Melford Cefuroxime Sodium Salt

Product Code. 30018177
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018177

Brand: Melford C563001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ceftriaxone Disodium Salt is a third-generation cephalosporin antibiotic. The hemi (heptahydrate) form indicates the presence of water molecules in the crystal structure.

Ceftriaxone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis. This interference leads to structural instability and eventual cell lysis. Broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. May be used in microbiology research and studies focusing on antibacterial agents. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.

Specifications

Chemical Name or Material Cefuroxime, Sodium Salt
CAS 56238-63-2
Quantity 1 g
Molecular Formula C16H15N4NaO8S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.