missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ DMEM/Ham's Medium F-12 Mix
BIOPHARMA

Product Code. 15363541
Change view
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15363541 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15363541 Supplier Corning™ Supplier No. 10080CV
  • ONLINE EXCLUSIVE PRICE
    161.00 €
    47.64 € / Pack of 6
    Save 113.36 €
    70% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos. Corning™ cellgro™ DMEM/Ham's Medium F-12 Mix is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.

  • Formulated for the serum-free culture of CHO cells
  • Formulation contains contains L-glutamine
  • Low levels of endotoxin, and absence of contamination
  • Osmolality: 289mOsm/kg H2O
  • Endotoxin: <0.25EU/mL
  • Sterility: Pass
  • Mycoplasma: Tested Negative

Order Info

dimQuantity

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 33.3mg/L CaCl2 (anhydrous), 0.0016mg/L CuSO2 (anhydrous), 0.834mg/L FeSO4.7H2O, 223.6mg/L KCl, 72.20mg/L MgSO4 (anhydrous), 7600mg/L NaCl, 142mg/L Na2HPO4 (anhydrous), 1176mg/L NaHCO3, 0.863mg/L ZnSO4.7H2O; Amino Acids: 8.9mg/L L-Alanine, 211mg/L L-Arginine. HCl, 15mg/L L-Asparagine.H2O, 13mg/L L-Aspartic acid, 35.12mg/L L-Cysteine.HCl.H2O, 14.70mg/L L-Glutamic acid, 146.20mg/L L-glutamine, 7.50mg/L Glycine, 20.96mg/L L-Histidine.HCl.H2O, 3.94mg/L L-Isoleucine, 13.10mg/L L-Leucine, 36.50mg/L L-Lysine. HCl, 4.48mg/L L-Methionine, 4.96mg/L L-Phenylalanine, 34.50mg/L L-Proline, 10.50mg/L L-Serine, 11.90mg/L L-Threonine, 2.04mg/L L-Tryptophan, 7.84mg/L L-Tyrosine.2Na.2H2O, 11.70mg/L L-Valine; Vitamins: 0.037mg/L Nicotinamide, 0.062mg/L Pyridoxine. HCl, 0.038mg/L Riboflavin, 0.34mg/L Thiamine. HCl, 1.36mg/L Vitamin B12; Other: 1802mg/L D-Glucose, 4.77mg/L Hypoxanthine, Na, 0.21mg/L DL-Thioctic (lipoic) acid, 0.088mg/L Methyl lineoleate, 1.2mg/L Phenol red. Na, 0.16mg/L Putrescine.2HCl, 110mg/L Sodium pyruvate, 0.73mg/L Thymidine.
pH 7.2
Content And Storage 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.