missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DMEM (Dulbecco's Modified Eagle's Medium)

Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
1 x 10 L
1 x 20 L
With Additives:
Glucose
Glucose, Glutamine
Glucose, Glutamine, Sodium Pyruvate
Glucose, Sodium Pyruvate
Glucose, Sodium Pyruvate, HEPES
Glutamine, Glucose
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
No Glutamine, No Sodium Pyruvate
No Sodium Pyruvate
None
Product Type:
DMEM Media, 1 g/L Glucose
DMEM Media, 4.5 g/L Glucose
missing translation for 'unitSize'
10 L
20 L
Pack of 6
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Original formulation was used to culture embryonic mouse cells
- Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
- Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
- Tested to determine pH and osmolality
- Low levels of endotoxin, and absence of contamination
- Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
- Osmolality: 335±mOsm/kg H2O
- Mycoplasma: Tested Negative
- Sterility: Pass
Specifications
Specifications
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM Media, 4.5 g/L Glucose |
| With Additives | Glutamine, Glucose |
| Without Additives | No Sodium Pyruvate |
| For Use With (Application) | To assess growth promotion and cytotoxicity characteristics |
| Quantity | 6 x 1 L |
| Endotoxin Level | <0.25EU/mL |
| Formulation | Inorganic Salts: 200mg/L CaCl2 (anhydrous), 0.1mg/L Fe(NO3)3.9H2O, 400mg/L KCl, 97.7mg/L MgSO4 (anhydrous), 6400mg/L NaCl, 125mg/L NaH2PO4.H2O, 3700mg/L NaHCO3; Amino Acids: 84mg/L L-Arginine. HCl, 62.57mg/L L-Cystine.2HCl, 584mg/L L-glutamine, 30mg/L Glycine, 42mg/L L-Histidine.HCl.H2O, 104.8mg/L L-Isoleucine, 104.8mg/L L-Leucine, 146.2mg/L L-Lysine. HCl, 30mg/L L-Methionine, 66mg/L L-Phenylalanine, 42mg/L L-Serine, 95.2mg/L L-Threonine, 16mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 20mg/L L-Valine; Vitamins: 4mg/L D-Calcium pantothenate, 4mg/L Choline chloride, 4mg/L Folic Acid, 7.2mg/L i-Inositol, 4mg/L Nicotinamide, 4mg/L Pyridoxine. HCl, 0.4mg/L Riboflavin, 4mg/L Thiamine. HCl; Other: 4500mg/L D-Glucose, 15mg/L Phenol red. Na. |
| Content And Storage | 2 to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction