missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ Improved MEM
BIOPHARMA

Product Code. 15393531
Change view
Click to view available options
With Additives:
L-Glutamine
Without Additives:
No Phenol Red
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. With Additives Without Additives Unit Size
15393531 L-Glutamine No Phenol Red Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15393531 Supplier Corning™ Supplier No. 10026CV
  • ONLINE EXCLUSIVE PRICE
    293.00 €
    107.81 € / Pack of 6
    Save 185.19 €
    63% Off
 Restricted Item
Estimated Shipment: 01-04-2027
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY

Especificaciones

Concentration 1 X
Product Type Improved MEM
With Additives L-Glutamine
Without Additives No Phenol Red
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6/Pk.
Endotoxin Level <0.25EU/mL
Formulation L-Glutamine
Content And Storage 2 to 8°C
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.