missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ MEM
BIOPHARMA

Product Code. 15333551
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15333551 500 mL Pack of 6
15383611 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15333551 Supplier Corning™ Supplier No. 15010CV
  • ONLINE EXCLUSIVE PRICE
    143.00 €
    131.10 € / Pack of 6
    Save 11.90 €
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Product Type Minimum Essential Medium (MEM)
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 500 mL
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 200.00mg/L CaCl2 (anhydrous), 400.00mg/L KCl, 97.70mg/L MgSO4 (anhydrous), 6,800.00mg/L NaCl, 140.00mg/L NaH2PO4.H2O, 2,200.00mg/L NaHCO3; Amino Acids: 126.40mg/L L-Arginine. HCl, 31.20mg/L L-Cystine.2HCl, 41.90mg/L L-Histidine.HCl.H2O, 52.50mg/L L-Isoleucine, 52.50mg/L L-Leucine, 72.50mg/L L-Lysine. HCl, 15.00mg/L L-Methionine, 32.50mg/L L-Phenylalanine, 47.60mg/L L-Threonine, 10.00mg/L L-Tryptophan, 51.90mg/L L-Tyrosine.2Na.2H2O, 46.80mg/L L-Valine; Vitamins: 1.00mg/L D-Calcium pantothenate, 1.00mg/L Choline chloride, 1.00mg/L Folic acid, 2.00mg/L i-Inositol, 1.00mg/L Nicotinamide, 1.00mg/L Pyridoxine. HCl, 0.10mg/L Riboflavin, 1.00mg/L Thiamine. HCl; Other: 1,000.00mg/L D-Glucose, 10.00mg/L Phenol red. Na.
Content And Storage 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.