Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Product Code. 10149081
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 40
Product Code. Color Unit Size
10149081 Black, Clear Bottom Pack of 40
1 options
Product Code. 10149081 Supplier Greiner Bio-One Supplier No. 781092
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
White, sterile microplate with lid.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-cytotoxic
  • Compatible with automated systems
  • Flexible working volume
  • Rounded square well profile
  • Stackable
  • Alphanumeric well cod
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 16 x 24
Packaging 10/Pk., 40/Cs.
Color Black, Clear Bottom
Product Type Clear Bottom
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
No. per Case 40
No. per Pack 10
Quantity 40/Cs.