Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Product Code. 10799404
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 32
Product Code. Color Unit Size
10799404 Black Pack of 32
1 options
Product Code. 10799404 Supplier Greiner Bio-One Supplier No. 655086
  • ONLINE EXCLUSIVE PRICE
    220.00 €
    187.64 € / Pack of 32
    Save 32.36 €
    14% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Color:Black; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Black
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Surface Treatment Tissue Culture
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 32
No. per Pack 8
Quantity 32/Cs.
Temperature Range (Metric) -20°C to +60°C
Well Design Chimney