missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry

CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry

Brand:  Invitrogen™ C34554

271.53 EUR valid until 2023-04-30
Use promo code "18437" to get your promotional price.



Additional Details : Weight : 0.00454kg

 View more versions of this product

Product Code. 10013162

  • 287.00 € / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CellTrace™ CFSE Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

  • Superior performance—bright, single-peak staining enables visualization of multiple generations
  • Long-term signal stability—well-retained in cells for several days post stain
  • Non-toxic—staining does not adversely effect cell health
  • Simple, robust staining protocol

    View a selection guide for all CellTrace™ Cell Proliferation Kits for flow cytometry.

    Superior fluorescent staining
    Successful proliferation analysis by dye dilution (see figure below) requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace™ CFSE dye enables the visualization of eight or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. Consistent, homogeneous staining results in very little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.

    Long-term signal retention
    Unlike stains that label the lipid membrane of cells, CellTrace™ CFSE dye easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye is designed to provide a consistent signal, even after several days in a cell culture environment.

    Non-toxic
    CellTrace™ CFSE dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. Researchers have used CFSE SE labeling to show that transplantable hematopoietic cells proliferate in vitro in response to stimulation by a growth factor cocktail.

    Simple, robust staining protocol
    The CellTrace™ CFSE Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
  • Specifications

    Specifications

    Contains 10 vials of CellTrace™ CFSE (lyophilized powder) and 1 vial of DMSO (500 μL). Store in freezer (-5 to -30°C) and protect from light.
    Fluorescent
    Flow Cytometer
    Other Label(s) or Dye(s)
    Lyophilized
    CellTrace™
    CFSE & Related Compounds
    DMSO (Dimethylsulfoxide)
    CellTrace™ Cell Proliferation Kit, 180 rxns, CFSE, Lyophilized, Fluorescent Detection Method, 492/517nm Excitation/Emission Wavelength, CellTrace™ CFSE Label/Dye, –5 to –30°C Storage, Use with Flow Cytometer
    Proliferation Assay
    Cell Proliferation Kit
    488
    Tube(s)
    1 Kit(s)
    Room Temperature
    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.