Learn More
Thermo Scientific Chemicals Cellulase, chromatographically purified, T. reesei
82.00 € - 212.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420337
|
Thermo Scientific Chemicals
J64019.MD |
250 mg |
82.00 €
250mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15416159
|
Thermo Scientific Chemicals
J64019.03 |
1 g |
212.00 €
1g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cellulase has been used to degrade cello-oligosaccharides into glucose and to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose. It is added to detergents to improve cleansing properties.