missing translation for 'onlineSavingsMsg'
Learn More

Cellulose Acetate Butyrate, Butyryl Content 50-54%

Product Code. p-84085 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10157270

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-36-8 | (C12H14O10)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-36-8
Molecular Formula (C12H14O10)n
MDL Number MFCD00081513

Specifications

Color White
Physical Form Granular Crystalline Powder
Melting Point 127.0°C to 240.0°C
Avg. Mol. Wt. or Mol. Wt. Range 16000 (Mn)
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ketones, organic acetates, lactates,, methylene, ethylene, propylene chlorides and, high-boiling solvents
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.475
Viscosity 0.01 to 0.03 (sec.)
Chemical Name or Material Cellulose acetate butyrate, Butyryl Content 50-54%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.