missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MDSYSAPEST PSASSRPEDY FIGATPLQKR LESVRKQSSF ILTPPRRKIP QCSQLQEDVD PQKVAFLLHK QWTLYSLTPL YKF Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 68%.
CENPL is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
Specifications
Specifications
| Antigen | CENPL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CENPL |
| Gene Accession No. | Q8N0S6 |
| Gene Alias | 2610300B10Rik; AW121806; AW550697; C1orf155; CENPL; CENP-L; Centromere protein L; dJ383J4.3; ICEN33; Interphase centromere complex protein 33; RGD1311868; RP3-383J4.1 |
| Gene Symbols | CENPL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?