missing translation for 'onlineSavingsMsg'
Learn More

Centralite II, TRC

Product Code. 30012238
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012238 250 mg 250mg
30011442 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012238 Supplier Toronto Research Chemicals Supplier No. TRCC256618250MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-92-7
Molecular Formula C15 H16 N2 O
Molecular Weight (g/mol) 240.31
Synonym N,N'-Dimethyl-N,N'-diphenylurea, 1,3-Dimethyl-1,3-diphenylurea, α,β-Dimethylcarbanilide, Dimethylcarbanilide, Methyl Centralite, N,N'-Dimethylcarbanilide, NSC 59781
IUPAC Name 1,3-dimethyl-1,3-diphenylurea
SMILES CN(C(=O)N(C)c1ccccc1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H16N2O/c1-16(13-9-5-3-6-10-13)15(18)17(2)14-11-7-4-8-12-14/h3-12H,1-2H3
Formula Weight 240.1263
Percent Purity >95%
Chemical Name or Material Centralite II
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.