missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP120 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 462.00 €
Specifications
| Antigen | CEP120 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CEP120 Polyclonal antibody specifically detects CEP120 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CEP120 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 153241 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CCDC100, centrosomal protein 120kDa, centrosomal protein of 120 kDa, coiled-coil domain containing 100, coiled-coil domain-containing protein 100, DKFZp686I06246, FLJ36090 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CEP120 (NP_694955.2). MVSKSDQLLIVVSILEGRHFPKRPKHMLVVEAKFDGEQLATDPVDHTDQPEFATELAWEIDRKALHQHRLQRTPIKLQCFALDPVTSAKETIGYIVLDLRTAQETKQAPKWYQLLSNKYTKFKSEIQISIALETDTKPPVDSFKAKGAPPRDGKVPAILAGLDPRDIVAVLNEEGGYHQIGPAEYCTDSFIMSVTIAFATQLEQLIPCTMKLPERQPEFFFYYSLLGNDVTNEPFNDLINPNFEPERASVRIRSSVEILRVYLALQSKLQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title