missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP120 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92822-0.1ml
This item is not returnable.
View return policy
Description
CEP120 Polyclonal antibody specifically detects CEP120 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CEP120 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCDC100, centrosomal protein 120kDa, centrosomal protein of 120 kDa, coiled-coil domain containing 100, coiled-coil domain-containing protein 100, DKFZp686I06246, FLJ36090 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CEP120 (NP_694955.2). MVSKSDQLLIVVSILEGRHFPKRPKHMLVVEAKFDGEQLATDPVDHTDQPEFATELAWEIDRKALHQHRLQRTPIKLQCFALDPVTSAKETIGYIVLDLRTAQETKQAPKWYQLLSNKYTKFKSEIQISIALETDTKPPVDSFKAKGAPPRDGKVPAILAGLDPRDIVAVLNEEGGYHQIGPAEYCTDSFIMSVTIAFATQLEQLIPCTMKLPERQPEFFFYYSLLGNDVTNEPFNDLINPNFEPERASVRIRSSVEILRVYLALQSKLQ | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 153241 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction