missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP135 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 135 kDa. Purity is >95% by SDS-PAGE.
LATS1 is a putative serine/threonine kinase that localizes to the mitotic apparatus and complexes with cell cycle controller CDC2 kinase in early mitosis. The protein is phosphorylated in a cell-cycle dependent manner, with late prophase phosphorylation remaining through metaphase. The N-terminal region of the protein binds CDC2 to form a complex showing reduced H1 histone kinase activity, indicating a role as a negative regulator of CDC2/cyclin A. In addition, the C-terminal kinase domain binds to its own N-terminal region, suggesting potential negative regulation through interference with complex formation via intramolecular binding. Biochemical and genetic data suggest a role as a tumor suppressor. This is supported by studies in knockout mice showing development of soft-tissue sarcomas, ovarian stromal cell tumors and a high sensitivity to carcinogenic treatments.
Specifications
Specifications
| Antigen | CEP135 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP135 |
| Gene Accession No. | Q66GS9 |
| Gene Alias | BC062951; centrosomal protein 135; centrosomal protein 135kDa; centrosomal protein 4; Centrosomal protein of 135 kDa; centrosomal protein of 135 kDa; LOW QUALITY PROTEIN: centrosomal protein of 135 kDa; centrosome protein cep135; Cep135; Cep4; Gm1044; KIAA0635; MCPH8; RGD1310726 |
| Gene Symbols | CEP135 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?