missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP152 Monoclonal Antibody (P1285)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518285
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
Specifications
| CEP152 | |
| Monoclonal | |
| 1.65 mg/mL | |
| PBS with no preservative | |
| O94986 | |
| CEP152 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human CEP152. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| P1285 | |
| Unconjugated | |
| CEP152 | |
| AI851464; asterless; CC-CKR-8; CCR-8; CDw198; centrosomal protein 152; centrosomal protein 152kDa; centrosomal protein of 152 kDa; CEP152; CKRL1; CMKBR8; CMKBRL2; CY6; GPRCY6; Kiaa0912; MCPH4; MCPH9; microcephaly, primary autosomal recessive 4; mKIAA0912; RGD1305348; SCKL5; TER1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 22995 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction