missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP170 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92228-0.1ml
This item is not returnable.
View return policy
Description
CEP170 Polyclonal antibody specifically detects CEP170 in Human samples. It is validated for Western Blot
Specifications
| CEP170 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| centrosomal protein 170kDa, Cep170, KARP 1 binding protein, KARP-1-binding protein, KARP1-binding protein, KIAA0470FAM68AKABcentrosomal protein of 170 kDa, XRCC5 binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1505-1584 of human CEP170 (NP_055627.2). NMGFPSAMLPSPPKQKSSPVNNHHSPGQTPTLGQPEARALHPAAVSAAAEFENAESEADFSIHFNRFNPDGEEEDVTVQE | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 9859 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction