missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP350 Antibody (CL3423) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59028PE
This item is not returnable.
View return policy
Description
CEP350 Monoclonal antibody specifically detects CEP350 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CEP350 | |
| Monoclonal | |
| PE | |
| CAP350GM133, centrosomal protein 350kDa, centrosome-associated protein 350, Centrosome-associated protein of 350 kDa, Cep350, FLJ38282, FLJ44058, KIAA0480centrosome associated protein 350 | |
| This antibody was developed using a recombinant protein derived from Q5VT06, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3423 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9857 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction