missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Cephalexin, Monohydrate
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Cephalexin Monohydrate is a cephalosporin antibiotic. Cephalexin, as a cephalosporin, inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
This interference disrupts the cross-linking of peptidoglycans in the bacterial cell wall, leading to structural instability and eventual cell lysis. Cephalexin is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalexin.
Specifications
Specifications
| Chemical Name or Material | Cephalexin |
| CAS | 23325-78-2 |
| Quantity | 10 g |
| Molecular Formula | C16H17N3O4S-H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction