missing translation for 'onlineSavingsMsg'
Learn More

Melford Cephalexin, Monohydrate

Product Code. 30018183
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018183 10 g 10g
30018184 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018183 Supplier Melford Supplier No. C5900010.0
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cephalexin Monohydrate is a cephalosporin antibiotic. Cephalexin, as a cephalosporin, inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).

This interference disrupts the cross-linking of peptidoglycans in the bacterial cell wall, leading to structural instability and eventual cell lysis. Cephalexin is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalexin.

Specifications

Chemical Name or Material Cephalexin
CAS 23325-78-2
Quantity 10 g
Molecular Formula C16H17N3O4S-H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.