missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin monohydrate, MedChemExpress

Product Code. 16492079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492079 1 g 1g
16430818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492079 Supplier Medchem Express Supplier No. HYB0200B
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cephalexin (Cefalexin) monohydrate is a potent, orally active new semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum.

Cephalexin (Cefalexin) monohydrate has antibacterial activity against a wide variety of gram-positive and gram-negative bacteria. Cephalexin (Cefalexin) monohydrate targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. Cephalexin (Cefalexin) monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, et al.

Specifications

Chemical Name or Material Cephalexin monohydrate
Physical Form Solid
CAS 23325-78-2
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C16H19N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym Cefalexin hydrate Cephacillin hydrate
Solubility Information DMSO : 6.67 mg/mL (18.25 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.47 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(C)CS[C@]2([H])[C@H](NC([C@H](N)C3=CC=CC=C3)=O)C1=O)O.O
Molecular Weight (g/mol) 365.4
Formula Weight 365.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.