missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Monohydrate USP MP Biomedicals

Product Code. 11417891 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417891 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417891 Supplier MP Biomedicals Supplier No. 0215058505
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23325-78-2 | Formula: C16H19N3O5S | Weight: 365.40 g/mol

Chemical Identifiers

CAS 23325-78-2
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.40
MDL Number MFCD00167148,MFCD00056877
InChI Key AVGYWQBCYZHHPN-CYJZLJNKSA-N
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate
PubChem CID 62921
ChEBI CHEBI:3535
IUPAC Name (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
SMILES O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O

Specifications

Type Cephalexin Monohydrate
Physical Form Crystalline Powder
Color Beige to White
Quantity 5 g
Formula Weight 365.4
Percent Purity ≥95%
Grade BP/USP
Assay ≥95%
Chemical Name or Material Cephalexin Monohydrate
compliance-icons
Product Identifier
  • Cephalexin Monohydrate BP, USP grade
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.