missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceramide Kinase Like Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92288-0.02ml
This item is not returnable.
View return policy
Description
Ceramide Kinase Like Polyclonal antibody specifically detects Ceramide Kinase Like in Mouse samples. It is validated for Western Blot
Specifications
| Ceramide Kinase Like | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ceramide kinase-like, ceramide kinase-like protein, retinitis pigmentosa 26 (autosomal recessive), RP26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 359-558 of human CERKL (NP_001025482.1). LAEKYRWMSPNQRRDFAVVKALAKLKAEDCEISFLPFNSSDDVQERRAQGSPKSDCNDQWQMIQGQFLNVSIMAIPCLCSVAPRGLAPNTRLNNGSMALIIARNTSRPEFIKHLKRYASVKNQFNFPFVETYTVEEVKVHPRNNTGGYNPEEEEDETASENCFPWNVDGDLMEVASEVHIRLHPRLISLYGGSMEEMIPK | |
| 0.02 mL | |
| Vision | |
| 375298 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction