missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cerebellin 4 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149242
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Cerebellin (CER), which was originally isolated from rat cerebellum, is a hexadecapeptide derived from a larger precursor called Cerebellin 1, also designated precerebellin 1 or Cbln1. Four propeptides, Cerebellin 1, Cerebellin 2 (Cbln2), Cerebellin 3 (Cbln3) and Cerebellin 4 (Cbln4), comprise the precerebellin subfamily within the C1q protein family. Cerebellin family members act as transneuronal regulators of synapse development and synaptic plasticity in various brain regions. Cerebellin and its metabolite, des-Ser(1)Cer, are also expressed in several extra-cerebellar tissues, including adrenal gland. Cerebellin 1, 2 and 3 assemble into homomeric and heteromeric complexes, thereby influencing each other's degradation and secretion. Cerebellin 3 is not able to form homomeric complexes, and can only be secreted upon forming a heteromeric complex with Cerebellin 1. Decreased concentrations of Cerebellin have been found in the brain of patients with olivopontocerebellar atrophy (OPCA) and Shy-Drager syndrome, suggesting a role for Cerebellin in the pathology of these diseases.
Specifications
| Cerebellin 4 | |
| Polyclonal | |
| Unconjugated | |
| CBLN4 | |
| AI848962; Cbln4; CBLNL1; cerebellin 4 precursor; cerebellin 4 precursor protein; cerebellin precursor-like 1; Cerebellin4; cerebellin-4; Cerebellin-like glycoprotein 1; UNQ718/PRO1382 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 140689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NTU7 | |
| CBLN4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Cerebellin-4, Gln28-Leu201. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction