missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)

Product Code. p-7059379
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11485200

Brand: Thermo Scientific Alfa Aesar 044496.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54451-25-1 | C3Ce2O9 | 460.26 g/mol

Cerium(III) carbonate hydrate is used to prepare cerium chloride and incandescent lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
PubChem CID 16212049
IUPAC Name dicerium(3+) tricarbonate
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 848°C
Physical Form Crystalline
Quantity 25 g
Merck Index 14,1996
Solubility Information Soluble in dilute mineral acids. Insoluble in water.
Formula Weight 460.27 (Anhydrous)
Percent Purity 99.999%
Grade Reacton
Assay (REO)
Chemical Name or Material Cerium (III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.