missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate hydrate, 98%

Product Code. p-7080565
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439412 50 g 50g
11449412 250 g 250g
11429412 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439412 Supplier Thermo Scientific Alfa Aesar Supplier No. A10142.18
Add to basket
This item is not returnable. View return policy

CAS: 95838-16-7 | CeO8S2 | 332.23 g/mol

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95838-16-7
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
PubChem CID 20223355
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 195°C (decomposition)
Quantity 50 g
UN Number UN3085
Merck Index 14,1990
Solubility Information Soluble in water.
IUPAC Name cerium(4+);disulfate;hydrate
Formula Weight 332.24 (Anhydrous)
Percent Purity 98%
Density 3.01
Chemical Name or Material Cerium (IV) sulfate hydrate
compliance-icons compliance-icons
Product Identifier
  • Cerium(IV) sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Cerium(IV) sulfate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.